Product Details

SNP ID
rs4491227
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:89670591 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CAAACAAACAAACAAACAAACAAAC[A/G]AAGAAAATCAGTGGAACTCACCTAA
Phenotype
MIM: 609020
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
FOLH1B PubMed Links

Gene Details

Gene
FOLH1B
Gene Name
folate hydrolase 1B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_153696.2 Intron NP_710163.1
XM_017017319.1 Intron XP_016872808.1

View Full Product Details