Product Details

SNP ID
hCV26861708
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:51542956 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AAAGTATACCAGGCAGCATTGTGTT[C/T]TATATGGCCCCTGATCAGGACATTT
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
RAD54L2 PubMed Links

Gene Details

Gene
RAD54L2
Gene Name
RAD54-like 2 (S. cerevisiae)
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001322253.1 Intron NP_001309182.1
NM_001322256.1 Intron NP_001309185.1
NM_001322257.1 Intron NP_001309186.1
NM_015106.3 Intron NP_055921.2

View Full Product Details