Product Details

SNP ID
rs36184181
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:56700469 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCCAGGAATGCAGCTGGGCCTCTTC[G/T]TGGTGTTCCTGGGCGTGTACTCTCT
Phenotype
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
OR9G1 PubMed Links

Gene Details

Gene
OR9G1
Gene Name
olfactory receptor family 9 subfamily G member 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001005213.1 82 Missense Mutation GTG,TTG V28L NP_001005213.1

View Full Product Details