Product Details

SNP ID
rs34194184
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:138617313 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTTGAATAGCACCCCTAAATACATT[A/C]TGATTTACAAAGGATGCCTTATCAA
Phenotype
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
FAIM PubMed Links

Gene Details

Gene
FAIM
Gene Name
Fas apoptotic inhibitory molecule
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001033030.1 Intron NP_001028202.1
NM_001033031.1 Intron NP_001028203.1
NM_001033032.1 Intron NP_001028204.1
NM_018147.3 Intron NP_060617.1
XM_011512950.2 Intron XP_011511252.1

View Full Product Details