Product Details

SNP ID
rs13099364
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:108303816 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACATAGTAGGTACGTGATGGGCTCT[C/T]GATTTTTTTTTCTCATTTTGTCTGA
Phenotype
MIM: 604371
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
HHLA2 PubMed Links

Gene Details

Gene
HHLA2
Gene Name
HERV-H LTR-associating 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001282556.1 Intron NP_001269485.1
NM_001282557.1 Intron NP_001269486.1
NM_001282558.1 Intron NP_001269487.1
NM_001282559.1 Intron NP_001269488.1
NM_007072.3 Intron NP_009003.1
XM_005247080.3 Intron XP_005247137.1
XM_011512362.1 Intron XP_011510664.1
XM_011512363.1 Intron XP_011510665.1
XM_011512364.1 Intron XP_011510666.1
XM_011512367.2 Intron XP_011510669.1
XM_017005642.1 Intron XP_016861131.1

View Full Product Details