Product Details

SNP ID
rs4724692
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:5470948 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGTCCTTGCCAATGGTATCTGAGTA[A/G]GGACTCCCCTCCCTCATCCGTGCAG
Phenotype
MIM: 609084
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
FBXL18 PubMed Links

Gene Details

Gene
FBXL18
Gene Name
F-box and leucine rich repeat protein 18
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001321213.1 Intron NP_001308142.1
NM_024963.5 Intron NP_079239.3
XM_011515531.2 Intron XP_011513833.1
XM_011515532.2 Intron XP_011513834.1

View Full Product Details