Product Details

SNP ID
rs9900341
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:1286250 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGTCGTTTGCTAAGAACTAGGGGCC[C/T]GGTGGCAGCAGGGAGGGGAGGTGAC
Phenotype
MIM: 612211
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
TUSC5 PubMed Links

Gene Details

Gene
TUSC5
Gene Name
tumor suppressor candidate 5
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_172367.2 Intron NP_758955.2

View Full Product Details