Product Details

SNP ID
rs1732805
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:113060959 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGCAGGGACAGACTCCCCACAAGGC[C/T]GCCAAGTTGGCAACTTTACCTTTGA
Phenotype
MIM: 602582
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
DTX1 PubMed Links

Gene Details

Gene
DTX1
Gene Name
deltex E3 ubiquitin ligase 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_004416.2 Intron NP_004407.2
XM_011538009.2 Intron XP_011536311.1

View Full Product Details