Product Details

SNP ID
rs11688605
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:28906552 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GTTCAGATGATAAACATATTGTGGA[A/G]TGGAACGTACAGACATGCAAAGTAA
Phenotype
MIM: 616195
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
SNORD92 PubMed Links

Gene Details

Gene
SNORD92
Gene Name
small nucleolar RNA, C/D box 92
There are no transcripts associated with this gene.

Gene
WDR43
Gene Name
WD repeat domain 43
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_015131.2 512 Silent Mutation GAA,GAG E152E NP_055946.1

View Full Product Details