Product Details
- SNP ID
-
rs35537698
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.2:3704115 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GGTCTGAGCGTGGTCAGGGCCGTGC[C/T]CCCCAGGTGTCCTCCCCAGGCTTCC
- Phenotype
-
MIM: 612396
- Polymorphism
- C/T, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
ALLC
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs192635091] are located under a probe and SNP(s) [rs144689929] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- ALLC
- Gene Name
- allantoicase
There are no transcripts associated with this gene.
- Gene
- DCDC2C
- Gene Name
- doublecortin domain containing 2C
View Full Product Details