Product Details

SNP ID
rs2078348
Assay Type
Functionally tested
NCBI dbSNP Submissions
44
Location
Chr.1:28236573 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TTGGGCGCCCCATCCCCCAACAGAA[C/T]TCCCGGGCCCCAATCCAGTATACCC
Phenotype
MIM: 614981
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ATPIF1 PubMed Links

Gene Details

Gene
ATPIF1
Gene Name
ATPase inhibitory factor 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_016311.4 393 Intron NP_057395.1
NM_178190.2 393 Intron NP_835497.1
NM_178191.2 393 UTR 3 NP_835498.1
Gene
DNAJC8
Gene Name
DnaJ heat shock protein family (Hsp40) member C8
There are no transcripts associated with this gene.

Gene
LOC105376896
Gene Name
uncharacterized LOC105376896
There are no transcripts associated with this gene.

View Full Product Details