Product Details

SNP ID
rs4145897
Assay Type
Functionally tested
NCBI dbSNP Submissions
29
Location
Chr.1:160543383 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACAAGTGCATGGCCCTTGGTGACAA[A/G]AAGTCTTGAATTCAAATACTGTCTT
Phenotype
MIM: 604513
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CD84 PubMed Links

Gene Details

Gene
CD84
Gene Name
CD84 molecule
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001184879.1 5629 UTR 3 NP_001171808.1
NM_001184881.1 5629 UTR 3 NP_001171810.1
NM_001184882.1 5629 UTR 3 NP_001171811.1
NM_003874.3 5629 UTR 3 NP_003865.1
XM_011510094.2 5629 UTR 3 XP_011508396.1
XM_011510095.2 5629 UTR 3 XP_011508397.1

View Full Product Details