Product Details

SNP ID
rs35618389
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:35697612 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGGGGCCCTAGGGCATGAAGGCACT[G/T]GGGGTTGGGGAGGGGACAGGGGATG
Phenotype
MIM: 186745 MIM: 190990
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
TLN1 PubMed Links

Gene Details

Gene
TLN1
Gene Name
talin 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_006289.3 8159 UTR 3 NP_006280.3
Gene
TPM2
Gene Name
tropomyosin 2 (beta)
There are no transcripts associated with this gene.

View Full Product Details