Product Details

SNP ID
rs1135830
Assay Type
DME
NCBI dbSNP Submissions
NA
Location
Chr.22:42127895 on Build GRCh38
Set Membership
DME Validated Inventoried
Context Sequence [VIC/FAM]
CAGGAGGCCCCAGGCCAGCGTGGTC[A/G]AGGTGGTCACCATCCCGGCAGAGAA
Phenotype
MIM: 124030
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CYP2D6 PubMed Links
Additional Information
The CYP2D6 gene exhibits copy number variation. Individuals may carry deletion alleles or extra copies of CYP2D6. CYP2D6 SNP genotyping assays run on samples lacking CYP2D6 genes will not amplify, homozygous samples having 1 or more gene copies typically cluster together, and heterozygous samples with more than 2 copies may run between the 2 copy heterozygous and homozygous genotype clusters. In addition, some CYP2D6 alleles contain CYP2D7 pseudogene sequences. For accurate CYP2D6 genotype analysis, copy number analysis must be done. For more information, refer to the PGx Experiments User Guide (Pub. # MAN0009612) Chapter 2 Copy Number Variation section.

Gene Details

Gene
CYP2D6
Gene Name
cytochrome P450 family 2 subfamily D member 6
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000106.5 869 Missense Mutation NP_000097.3
NM_001025161.2 869 Missense Mutation NP_001020332.2
XM_011529966.2 869 Intron XP_011528268.1
XM_011529968.2 869 Intron XP_011528270.1
XM_011529970.2 869 Intron XP_011528272.1
XM_011529972.2 869 Intron XP_011528274.1
Gene
LOC102723722
Gene Name
uncharacterized LOC102723722
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
XM_017029174.1 869 Intron XP_016884663.1
Gene
NDUFA6-AS1
Gene Name
NDUFA6 antisense RNA 1 (head to head)
There are no transcripts associated with this gene.

View Full Product Details