Product Details

SNP ID
rs2957485
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:34109286 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATTGTCTTTCCTCTGCTGGCCACCC[A/C]ATACCCTTTGGAAACTTTCTCCTTC
Phenotype
MIM: 601178 MIM: 609221
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
CAPRIN1 PubMed Links

Gene Details

Gene
CAPRIN1
Gene Name
cell cycle associated protein 1
There are no transcripts associated with this gene.

Gene
NAT10
Gene Name
N-acetyltransferase 10
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001144030.1 Intron NP_001137502.1
NM_024662.2 Intron NP_078938.2

View Full Product Details