Product Details
- SNP ID
-
rs615709
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.11:64942329 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- TTGGGGAGTGAGTGGTGTCTGGTTA[C/T]ATGAATGAGTTTTTTTAATAGTGAT
- Phenotype
-
MIM: 609651
MIM: 601644
- Polymorphism
- C/T, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
GPHA2
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs112515366] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- GPHA2
- Gene Name
- glycoprotein hormone alpha 2
There are no transcripts associated with this gene.
- Gene
- MAJIN
- Gene Name
- membrane anchored junction protein
- Gene
- PPP2R5B
- Gene Name
- protein phosphatase 2 regulatory subunit B'beta
There are no transcripts associated with this gene.
View Full Product Details