Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_001080503.2 | Intron | NP_001073972.2 | |||
XM_006722643.1 | Intron | XP_006722706.1 | |||
XM_011527678.2 | Intron | XP_011525980.2 | |||
XM_017026255.1 | Intron | XP_016881744.1 | |||
XM_017026256.1 | Intron | XP_016881745.1 | |||
XM_017026257.1 | Intron | XP_016881746.1 | |||
XM_017026258.1 | Intron | XP_016881747.1 | |||
XM_017026259.1 | Intron | XP_016881748.1 | |||
XM_017026260.1 | Intron | XP_016881749.1 | |||
XM_017026261.1 | Intron | XP_016881750.1 | |||
XM_017026262.1 | Intron | XP_016881751.1 | |||
XM_017026263.1 | Intron | XP_016881752.1 | |||
XM_017026264.1 | Intron | XP_016881753.1 |
Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_001170635.1 | Intron | NP_001164106.1 | |||
NM_022737.2 | Intron | NP_073574.2 | |||
XM_005260017.1 | Intron | XP_005260074.1 | |||
XM_017027147.1 | Intron | XP_016882636.1 | |||
XM_017027148.1 | Intron | XP_016882637.1 |