Product Details

SNP ID
rs759682686
Assay Type
Functionally Tested
NCBI dbSNP Submissions
2
Location
Chr.16:1229723 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACCTGCCCCAGCCCCAGGCCAGGCC[C/G]TGCAGCAAACGGGCATTGTTGGGGG
Phenotype
MIM: 607904 MIM: 191081 MIM: 609341
Polymorphism
C/G, Transversion Substitution
Allele Nomenclature
Literature Links
CACNA1H PubMed Links

Gene Details

Gene
CACNA1H
Gene Name
calcium voltage-gated channel subunit alpha1 H
There are no transcripts associated with this gene.

Gene
TPSB2
Gene Name
tryptase beta 2 (gene/pseudogene)
There are no transcripts associated with this gene.

Gene
TPSG1
Gene Name
tryptase gamma 1
There are no transcripts associated with this gene.

View Full Product Details