Product Details

SNP ID
rs4272345
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.8:93709424 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TAATTTTTGATAAATGGCACAGGTA[A/G]TGAAATAGAGGGAAGCAGTAGAAAA
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
FAM92A1 PubMed Links

Gene Details

Gene
FAM92A1
Gene Name
family with sequence similarity 92 member A1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001283034.1 Intron NP_001269963.1
NM_145269.4 Intron NP_660312.2
XM_005250787.2 Intron XP_005250844.1
Gene
LINC00535
Gene Name
long intergenic non-protein coding RNA 535
There are no transcripts associated with this gene.

View Full Product Details