Product Details

SNP ID
rs4266953
Assay Type
Functionally tested
NCBI dbSNP Submissions
2
Location
Chr.1:212997611 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGCCTCCACTTACTAAAGAACATGT[A/G]TTCTAACAAGTTTTATTGTGTTTTT
Phenotype
MIM: 610471
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ANGEL2 PubMed Links

Gene Details

Gene
ANGEL2
Gene Name
angel homolog 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001300753.1 Intron NP_001287682.1
NM_001300755.1 Intron NP_001287684.1
NM_001300757.1 Intron NP_001287686.1
NM_001300758.1 Intron NP_001287687.1
NM_144567.4 Intron NP_653168.2
Gene
VASH2
Gene Name
vasohibin 2
There are no transcripts associated with this gene.

View Full Product Details