Product Details

SNP ID
rs12859195
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:23344059 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TGCTAAAATACAGACTGATTCAAAG[A/G]GTCTGGGGTGGGAACTGCAAGTCTG
Phenotype
MIM: 300828
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
PTCHD1 PubMed Links

Gene Details

Gene
PTCHD1
Gene Name
patched domain containing 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_173495.2 Intron NP_775766.2
XM_011545449.2 Intron XP_011543751.1

View Full Product Details