Product Details

SNP ID
rs10948227
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:45338370 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AAAAAAAAAAAATTATAGGCAACTT[C/G]TCAAGAAATTCTAAAATCAGTAATA
Phenotype
MIM: 600211 MIM: 602947
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
RUNX2 PubMed Links

Gene Details

Gene
RUNX2
Gene Name
runt related transcription factor 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001015051.3 Intron NP_001015051.3
NM_001024630.3 Intron NP_001019801.3
NM_001278478.1 Intron NP_001265407.1
XM_006715232.1 Intron XP_006715295.1
XM_011514960.2 Intron XP_011513262.1
XM_011514961.2 Intron XP_011513263.1
XM_011514962.2 Intron XP_011513264.1
XM_011514963.2 Intron XP_011513265.1
XM_011514964.2 Intron XP_011513266.1
XM_011514965.2 Intron XP_011513267.1
XM_011514966.2 Intron XP_011513268.1
XM_017011391.1 Intron XP_016866880.1
XM_017011392.1 Intron XP_016866881.1
XM_017011393.1 Intron XP_016866882.1
XM_017011394.1 Intron XP_016866883.1
XM_017011395.1 Intron XP_016866884.1
XM_017011396.1 Intron XP_016866885.1
Gene
SUPT3H
Gene Name
SPT3 homolog, SAGA and STAGA complex component
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001261823.1 Intron NP_001248752.1
NM_003599.3 Intron NP_003590.1
NM_181356.2 Intron NP_852001.1
XM_011514949.2 Intron XP_011513251.1
XM_011514950.1 Intron XP_011513252.1
XM_011514951.2 Intron XP_011513253.1
XM_011514952.2 Intron XP_011513254.1
XM_011514953.2 Intron XP_011513255.1
XM_011514954.2 Intron XP_011513256.1
XM_017011369.1 Intron XP_016866858.1
XM_017011370.1 Intron XP_016866859.1
XM_017011371.1 Intron XP_016866860.1
XM_017011372.1 Intron XP_016866861.1
XM_017011373.1 Intron XP_016866862.1
XM_017011374.1 Intron XP_016866863.1

View Full Product Details