Product Details
- SNP ID
-
rs270202
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.5:5141181 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CCTTTCCTCCTTCTCCTCCTTTATA[A/G]ATTAAAATATTCCTCTATCGTTGCT
- Phenotype
-
MIM: 607510
- Polymorphism
- A/G, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
ADAMTS16
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs116720952] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- ADAMTS16
- Gene Name
- ADAM metallopeptidase with thrombospondin type 1 motif 16
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_139056.3 |
|
Intron |
|
|
NP_620687.2 |
- Gene
- CTD-2297D10.2
- Gene Name
- uncharacterized LOC101929176
There are no transcripts associated with this gene.
View Full Product Details