Product Details

SNP ID
rs144688
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.21:10595541 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GCTGTCAGGAACAAGAGAAGAGGGA[T/C]GGGAAACCTGCTACTGAGGTTTGGA
Phenotype
MIM: 604336
Polymorphism
T/C, Transition substitution
Allele Nomenclature
Literature Links
TPTE PubMed Links

Gene Details

Gene
TPTE
Gene Name
transmembrane phosphatase with tensin homology
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001290224.1 Intron NP_001277153.1
NM_199259.3 Intron NP_954868.1
NM_199260.3 Intron NP_954869.1
NM_199261.3 Intron NP_954870.2

View Full Product Details