Product Details
- SNP ID
-
rs4143370
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.21:25564661 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TATCACTTGGACTGGGCACCAGCTA[C/G]TGCCCTTAACTAGCTGGTGTCTCCA
- Phenotype
-
MIM: 609337
- Polymorphism
- C/G, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
MIR155
PubMed Links
Gene Details
- Gene
- MIR155
- Gene Name
- microRNA 155
There are no transcripts associated with this gene.
- Gene
- MIR155HG
- Gene Name
- MIR155 host gene
There are no transcripts associated with this gene.
View Full Product Details