Product Details
- SNP ID
-
rs214777
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.20:2299955 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- TACTAAAAATACAAAAATTAGTGGG[A/G]CATGCTGGCACGCACTTGTAGTCCC
- Phenotype
-
MIM: 600238
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
TGM3
PubMed Links
Gene Details
- Gene
- TGM3
- Gene Name
- transglutaminase 3
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_003245.3 |
|
Intron |
|
|
NP_003236.3 |
View Full Product Details