Product Details
- SNP ID
-
rs13194997
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.6:158168576 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GGCGGGGCCGCACGGCAGTCCCCCC[C/T]TCGTTTAATCGTGGGTTTCCTGTGG
- Phenotype
-
MIM: 608780
MIM: 614725
- Polymorphism
- C/T, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
GTF2H5
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs148023066] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- GTF2H5
- Gene Name
- general transcription factor IIH subunit 5
- Gene
- SERAC1
- Gene Name
- serine active site containing 1
View Full Product Details