Product Details

SNP ID
rs3890167
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:8241781 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TATCATTATATCCCCCATTAATAAG[A/G]AAACAATAGCACAGAGAGATGCAGT
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
FAM86FP PubMed Links

Gene Details

Gene
FAM86FP
Gene Name
family with sequence similarity 86, member A pseudogene
There are no transcripts associated with this gene.

Gene
LOC101927905
Gene Name
uncharacterized LOC101927905
There are no transcripts associated with this gene.

View Full Product Details