Product Details

SNP ID
rs3092970
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:218163065 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CAGAAGATGATGCTCACCTCAGGGT[G/T]AAGCTGAGACTCCCTGGAGGAGCAT
Phenotype
MIM: 146929
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
CXCR1 PubMed Links

Gene Details

Gene
CXCR1
Gene Name
C-X-C motif chemokine receptor 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000634.2 2266 UTR 3 NP_000625.1

View Full Product Details