Product Details

SNP ID
rs3095297
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:31115952 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGCTAACCCTATGCCTGGGCACTGG[A/C]CTTCTCCCATATGGGATATAGTGTA
Phenotype
MIM: 611401 MIM: 602593 MIM: 613525
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
C6orf15 PubMed Links

Gene Details

Gene
C6orf15
Gene Name
chromosome 6 open reading frame 15
There are no transcripts associated with this gene.

Gene
CDSN
Gene Name
corneodesmosin
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001264.4 1719 UTR 3 NP_001255.3
Gene
PSORS1C1
Gene Name
psoriasis susceptibility 1 candidate 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_014068.2 1719 Intron NP_054787.2

View Full Product Details