Product Details

SNP ID
rs3096308
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:89273277 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTGTTAGAGGCACGAGCTGGTCCTC[C/G]TCATTTTTGCTCTGTACAGAGCTGT
Phenotype
MIM: 611192
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
ANKRD11 PubMed Links

Gene Details

Gene
ANKRD11
Gene Name
ankyrin repeat domain 11
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001256182.1 Intron NP_001243111.1
NM_001256183.1 Intron NP_001243112.1
NM_013275.5 Intron NP_037407.4
XM_006721181.1 Intron XP_006721244.1
XM_006721184.2 Intron XP_006721247.1
XM_011523051.2 Intron XP_011521353.1
XM_011523053.2 Intron XP_011521355.1
XM_011523054.2 Intron XP_011521356.1
XM_011523055.2 Intron XP_011521357.1
XM_011523056.2 Intron XP_011521358.1
XM_011523057.2 Intron XP_011521359.1
XM_017023182.1 Intron XP_016878671.1
XM_017023183.1 Intron XP_016878672.1
XM_017023184.1 Intron XP_016878673.1
XM_017023185.1 Intron XP_016878674.1
XM_017023186.1 Intron XP_016878675.1
XM_017023187.1 Intron XP_016878676.1

View Full Product Details