Product Details

SNP ID
rs3105153
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:55115410 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AAATGTTTCGGAGATGAGGCAACAA[C/T]AGGCAGTACTTCTCAGATTTATCTG
Phenotype
MIM: 140050
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CDC20B PubMed Links

Gene Details

Gene
CDC20B
Gene Name
cell division cycle 20B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001145734.2 Intron NP_001139206.2
NM_001170402.1 Intron NP_001163873.1
NM_152623.2 Intron NP_689836.2
XM_011543218.2 Intron XP_011541520.1
Gene
GZMA
Gene Name
granzyme A
There are no transcripts associated with this gene.

View Full Product Details