Product Details

SNP ID
rs3177295
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:10227542 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TATGCCCACACTGTGTCTATCCCCT[C/T]CCCCGTGACGTGGTCTGGAGTCCAA
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
FAM173B PubMed Links

Gene Details

Gene
FAM173B
Gene Name
family with sequence similarity 173 member B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001258388.1 640 Missense Mutation AAG,GAG K184E NP_001245317.1
NM_001258389.1 640 UTR 3 NP_001245318.1
NM_199133.3 640 Missense Mutation AAG,GAG K201E NP_954584.2
XM_011513964.1 640 UTR 3 XP_011512266.1

View Full Product Details