Product Details

SNP ID
rs3135695
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:74236235 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CAGCGTCTGGAGGCAGGCCTTTTTG[A/C]CTTTGTTTTGGCATCTTGGGGTGAA
Phenotype
MIM: 605719 MIM: 600404
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
LAT2 PubMed Links

Gene Details

Gene
LAT2
Gene Name
linker for activation of T-cells family member 2
There are no transcripts associated with this gene.

Gene
RFC2
Gene Name
replication factor C subunit 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001278791.1 Intron NP_001265720.1
NM_001278792.1 Intron NP_001265721.1
NM_001278793.1 Intron NP_001265722.1
NM_002914.4 Intron NP_002905.2
NM_181471.2 Intron NP_852136.1
XM_006716080.2 Intron XP_006716143.1
XM_017012492.1 Intron XP_016867981.1
XM_017012493.1 Intron XP_016867982.1

View Full Product Details