Product Details
- SNP ID
-
rs3181004
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.19:4795555 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TTAAGTCATTAAAAAGATACTCAAA[A/G]AGAATGGCCTGCCTGATGCGTCCTT
- Phenotype
-
MIM: 613538
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
FEM1A
PubMed Links
Gene Details
- Gene
- FEM1A
- Gene Name
- fem-1 homolog A
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_018708.2 |
|
Intron |
|
|
NP_061178.1 |
View Full Product Details