Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_001206736.1 | 3023 | UTR 3 | NP_001193665.1 | ||
NM_016143.4 | 3023 | UTR 3 | NP_057227.2 | ||
NM_018839.4 | 3023 | UTR 3 | NP_061327.2 | ||
XM_006723592.2 | 3023 | UTR 3 | XP_006723655.1 | ||
XM_006723593.2 | 3023 | UTR 3 | XP_006723656.1 | ||
XM_006723594.2 | 3023 | UTR 3 | XP_006723657.1 | ||
XM_011529300.1 | 3023 | UTR 3 | XP_011527602.1 | ||
XM_011529301.1 | 3023 | UTR 3 | XP_011527603.1 | ||
XM_011529302.2 | 3023 | UTR 3 | XP_011527604.1 | ||
XM_011529304.1 | 3023 | UTR 3 | XP_011527606.1 | ||
XM_011529305.2 | 3023 | Intron | XP_011527607.1 | ||
XM_017027956.1 | 3023 | UTR 3 | XP_016883445.1 | ||
XM_017027957.1 | 3023 | UTR 3 | XP_016883446.1 | ||
XM_017027958.1 | 3023 | UTR 3 | XP_016883447.1 |