Product Details

SNP ID
rs3186051
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:151364459 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TCAAGTTTTTTCCCAAACAGCAATT[A/T]GATTTTTTGTCCTGATTTCTCTTTG
Phenotype
MIM: 604698 MIM: 616595
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
AKAP12 PubMed Links

Gene Details

Gene
AKAP12
Gene Name
A-kinase anchoring protein 12
There are no transcripts associated with this gene.

Gene
ZBTB2
Gene Name
zinc finger and BTB domain containing 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_020861.2 2982 UTR 3 NP_065912.1
XM_005267076.2 2982 UTR 3 XP_005267133.1
XM_011536004.2 2982 UTR 3 XP_011534306.1
XM_017011137.1 2982 UTR 3 XP_016866626.1

View Full Product Details