Product Details

SNP ID
rs3192448
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:76137304 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGTGGACCTGTGTTGGGGGGCAGTT[C/T]TGGACCCTGACTTGGGCACTGTCCA
Phenotype
MIM: 602291
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
FOXJ1 PubMed Links

Gene Details

Gene
FOXJ1
Gene Name
forkhead box J1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001454.3 1670 UTR 3 NP_001445.2
Gene
RNF157
Gene Name
ring finger protein 157
There are no transcripts associated with this gene.

Gene
RNF157-AS1
Gene Name
RNF157 antisense RNA 1
There are no transcripts associated with this gene.

View Full Product Details