Product Details

SNP ID
rs3177524
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:18257105 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AACACATCACCCAAGATCTAGGCTG[G/T]GGAGGACTTTATTCATTAGACCCAG
Phenotype
MIM: 600128
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
KIAA1683 PubMed Links

Gene Details

Gene
KIAA1683
Gene Name
KIAA1683
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001145304.1 4395 UTR 3 NP_001138776.1
NM_001145305.1 4395 UTR 3 NP_001138777.1
NM_025249.3 4395 UTR 3 NP_079525.1
XM_005260084.1 4395 UTR 3 XP_005260141.1
XM_011528324.1 4395 UTR 3 XP_011526626.1
Gene
LOC729966
Gene Name
uncharacterized LOC729966
There are no transcripts associated with this gene.

Gene
PDE4C
Gene Name
phosphodiesterase 4C
There are no transcripts associated with this gene.

View Full Product Details