Product Details

SNP ID
rs3200205
Assay Type
Functionally tested
NCBI dbSNP Submissions
11
Location
Chr.1:35178051 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATCATTCTTACCTAAAGAATAATAA[A/G]AAAAAGTTAATATAAAAGACAAGGG
Phenotype
MIM: 605199
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
SFPQ PubMed Links

Gene Details

Gene
SFPQ
Gene Name
splicing factor proline and glutamine rich
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_005066.2 8641 Intron NP_005057.1
XM_005271112.4 8641 Intron XP_005271169.1
XM_005271113.4 8641 Intron XP_005271170.1
XM_005271115.4 8641 Intron XP_005271172.1
XM_011541950.2 8641 UTR 3 XP_011540252.1
XM_017002053.1 8641 UTR 3 XP_016857542.1
XM_017002054.1 8641 UTR 3 XP_016857543.1

View Full Product Details