Product Details
- SNP ID
-
rs3193507
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.11:74333931 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- AGAAAAAGATAAACATTGTAGTGCC[C/T]GAGGCTGAGACCTCCTAGCCAGCAC
- Phenotype
-
MIM: 611610
- Polymorphism
- C/T, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
LOC101928580
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs200287727,rs518268] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- LOC101928580
- Gene Name
- uncharacterized LOC101928580
There are no transcripts associated with this gene.
- Gene
- PGM2L1
- Gene Name
- phosphoglucomutase 2 like 1
View Full Product Details