Product Details

SNP ID
rs3210288
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.Y:7377198 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACACAGCATAAACTGAGGGTGTGGG[T/G]TCTGGCTTTGTACATAACTCATTTT
Phenotype
MIM: 400008
Polymorphism
T/G, Transversion substitution
Allele Nomenclature
Literature Links
PRKY PubMed Links

Gene Details

Gene
PRKY
Gene Name
protein kinase, Y-linked, pseudogene
There are no transcripts associated with this gene.

View Full Product Details