Product Details

SNP ID
rs3739775
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:70529413 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CCCCATTATAAACTAGGGTTAGAAC[A/G]TTTACAAATGAGAGTCTAGGTCCTA
Phenotype
MIM: 608961
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
TRPM3 PubMed Links

Gene Details

Gene
TRPM3
Gene Name
transient receptor potential cation channel subfamily M member 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001007470.1 14768 Intron NP_001007471.1
NM_001007471.2 14768 Intron NP_001007472.2
NM_020952.4 14768 Intron NP_066003.3
NM_024971.5 14768 Intron NP_079247.5
NM_206944.3 14768 Intron NP_996827.3
NM_206945.3 14768 Intron NP_996828.3
NM_206946.3 14768 Intron NP_996829.3
NM_206947.3 14768 Intron NP_996830.3
NM_206948.2 14768 Intron NP_996831.1
XM_011519035.2 14768 UTR 3 XP_011517337.1
XM_011519036.2 14768 UTR 3 XP_011517338.1
XM_011519037.2 14768 UTR 3 XP_011517339.1
XM_011519038.2 14768 UTR 3 XP_011517340.1
XM_011519039.2 14768 UTR 3 XP_011517341.1
XM_011519040.2 14768 UTR 3 XP_011517342.1
XM_011519041.2 14768 UTR 3 XP_011517343.1
XM_011519042.2 14768 UTR 3 XP_011517344.1
XM_011519043.2 14768 UTR 3 XP_011517345.1
XM_011519044.2 14768 UTR 3 XP_011517346.1
XM_011519045.2 14768 UTR 3 XP_011517347.1
XM_011519046.2 14768 UTR 3 XP_011517348.1
XM_011519047.2 14768 UTR 3 XP_011517349.1
XM_017015141.1 14768 UTR 3 XP_016870630.1
XM_017015142.1 14768 UTR 3 XP_016870631.1
XM_017015143.1 14768 UTR 3 XP_016870632.1
XM_017015144.1 14768 UTR 3 XP_016870633.1
XM_017015145.1 14768 UTR 3 XP_016870634.1
XM_017015146.1 14768 UTR 3 XP_016870635.1
XM_017015147.1 14768 UTR 3 XP_016870636.1
XM_017015148.1 14768 UTR 3 XP_016870637.1
XM_017015149.1 14768 UTR 3 XP_016870638.1
XM_017015150.1 14768 UTR 3 XP_016870639.1
XM_017015151.1 14768 UTR 3 XP_016870640.1
XM_017015152.1 14768 UTR 3 XP_016870641.1
XM_017015153.1 14768 UTR 3 XP_016870642.1
XM_017015154.1 14768 UTR 3 XP_016870643.1
XM_017015155.1 14768 UTR 3 XP_016870644.1
XM_017015156.1 14768 UTR 3 XP_016870645.1
XM_017015157.1 14768 UTR 3 XP_016870646.1
XM_017015158.1 14768 Intron XP_016870647.1
XM_017015159.1 14768 UTR 3 XP_016870648.1
XM_017015160.1 14768 UTR 3 XP_016870649.1
XM_017015161.1 14768 Intron XP_016870650.1

View Full Product Details