Product Details

SNP ID
rs3740865
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:94379871 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TTGAATGATTCAGCCCCCATCTGGG[C/T]CAACGTTGTCCTCGCTCCTCTTCCT
Phenotype
MIM: 605569
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
GPR83 PubMed Links

Gene Details

Gene
GPR83
Gene Name
G protein-coupled receptor 83
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_016540.3 1656 UTR 3 NP_057624.3
XM_011542559.2 1656 UTR 3 XP_011540861.1

View Full Product Details