Product Details

SNP ID
rs3731871
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:218640358 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTCTGTGTAAGAGCCAAGCAAATCT[A/G]CCCTTAGTCATACAAGTTTGAGTTG
Phenotype
MIM: 605939 MIM: 604083
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
PLCD4 PubMed Links

Gene Details

Gene
PLCD4
Gene Name
phospholipase C delta 4
There are no transcripts associated with this gene.

Gene
ZNF142
Gene Name
zinc finger protein 142
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001105537.2 Intron NP_001099007.1
XM_006712730.3 Intron XP_006712793.1
XM_011511784.2 Intron XP_011510086.1
XM_011511785.2 Intron XP_011510087.1
XM_011511786.2 Intron XP_011510088.1
XM_011511787.2 Intron XP_011510089.1
XM_011511788.2 Intron XP_011510090.1
XM_011511789.2 Intron XP_011510091.1
XM_017004871.1 Intron XP_016860360.1
XM_017004872.1 Intron XP_016860361.1

View Full Product Details