Product Details
- SNP ID
-
rs3747844
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.9:121463201 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- ACCCCTCCGACTTTGGCCGAACACC[A/G]TGGCTGGCTGCCGTGTGAAGAATGG
- Phenotype
-
MIM: 104175
- Polymorphism
- A/G, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
GGTA1P
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs77378583] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- GGTA1P
- Gene Name
- glycoprotein, alpha-galactosyltransferase 1 pseudogene
There are no transcripts associated with this gene.
View Full Product Details