Product Details

SNP ID
rs3752340
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:102140648 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AAAAATATCAGCTTAAGGTTAAAGC[A/G]TTGGGCCTGGCAAATATCTACCAGG
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
TCEAL6 PubMed Links

Gene Details

Gene
TCEAL6
Gene Name
transcription elongation factor A like 6
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001006938.2 845 UTR 3 NP_001006939.2

View Full Product Details