Product Details

SNP ID
rs3738455
Assay Type
Functionally tested
NCBI dbSNP Submissions
28
Location
Chr.1:25961798 on Build GRCh38
Set Membership
HapMap JSNP
Context Sequence [VIC/FAM]
CTGTTTGTCAATCAGCAAGGGATCC[C/T]AGTCCCAAAGTGATTTTAAGATCAA
Phenotype
MIM: 602344
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
PAFAH2 PubMed Links

Gene Details

Gene
PAFAH2
Gene Name
platelet activating factor acetylhydrolase 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000437.3 1303 UTR 3 NP_000428.2
XM_006710669.3 1303 UTR 3 XP_006710732.1
XM_006710670.3 1303 UTR 3 XP_006710733.1
XM_011541528.2 1303 UTR 3 XP_011539830.1
XM_011541529.2 1303 UTR 3 XP_011539831.1
XM_017001390.1 1303 UTR 3 XP_016856879.1
XM_017001391.1 1303 UTR 3 XP_016856880.1
XM_017001392.1 1303 UTR 3 XP_016856881.1

View Full Product Details