Product Details

SNP ID
rs3743012
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.15:42210609 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AAAAATCTTGGGAAGGCAAAATTGG[A/G]TAAAACCATTAAAACAGAAATAGAG
Phenotype
MIM: 612188
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
TMEM87A PubMed Links

Gene Details

Gene
TMEM87A
Gene Name
transmembrane protein 87A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001110503.2 2807 Intron NP_001103973.1
NM_001286487.1 2807 UTR 3 NP_001273416.1
NM_015497.4 2807 UTR 3 NP_056312.2
XM_005254287.1 2807 UTR 3 XP_005254344.1
XM_006720468.2 2807 Intron XP_006720531.1
XM_011521443.2 2807 Intron XP_011519745.1
XM_011521444.2 2807 Intron XP_011519746.1
XM_017022067.1 2807 Intron XP_016877556.1
Gene
VPS39
Gene Name
VPS39, HOPS complex subunit
There are no transcripts associated with this gene.

View Full Product Details