Product Details

SNP ID
rs3760912
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:13152524 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TGAAGGAAGCAGATAGAACAGTATC[C/T]TCCGAGGAGTGGAGGCTGGGGATCG
Phenotype
MIM: 603765
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
IER2 PubMed Links

Gene Details

Gene
IER2
Gene Name
immediate early response 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_004907.2 Intron NP_004898.2
Gene
STX10
Gene Name
syntaxin 10
There are no transcripts associated with this gene.

View Full Product Details